Assay Details
| Gene Symbol: RPL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243345.1 | NP_001230274.1 | 5 - 6 | 557 | 58 | |
| XM_013994186.2 | XP_013849640.1 | 713 | 58 | |||
| GenBank mRNA | AJ945945.1 | - | 2 - 3 | 233 | 58 | |
| AK231738.1 | - | 4 - 5 | 715 | 58 | ||
| AK237333.1 | - | 5 - 6 | 530 | 58 | ||
| AK347687.1 | - | 5 - 6 | 523 | 58 | ||
| AK350211.1 | - | 4 - 5 | 710 | 58 | ||
| BP171914.1 | - | 5 - 6 | 560 | 58 | ||
More Information
Set Membership: |
|

RPL13
Genomic Map