Assay Details
Gene Symbol: RANGRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243369.1 | NP_001230298.1 | 1 - 2 | 189 | 90 | |
NM_001243370.1 | NP_001230299.1 | 1 - 2 | 188 | 90 | ||
XM_005657038.3 | XP_005657095.1 | 225 | 90 | |||
XM_021066166.1 | XP_020921825.1 | 886 | 90 | |||
GenBank mRNA | AK232566.1 | - | 1 - 2 | 186 | 90 | |
AY609564.1 | - | 1 - 2 | 173 | 90 | ||
CK455001.1 | - | 1 - 2 | 188 | 90 | ||
FD587652.1 | - | 1 - 2 | 197 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005657038.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
