Assay Details
Gene Symbol: RANGRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243369.1 | NP_001230298.1 | 2 - 3 | 311 | 57 | |
NM_001243370.1 | NP_001230299.1 | 2 - 3 | 310 | 57 | ||
XM_021066166.1 | XP_020921825.1 | 1008 | 57 | |||
GenBank mRNA | AK232566.1 | - | 2 - 3 | 308 | 57 | |
AY609564.1 | - | 2 - 3 | 295 | 57 | ||
CK455001.1 | - | 2 - 3 | 310 | 57 | ||
FD587652.1 | - | 2 - 3 | 319 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
