Assay Details
Gene Symbol: RANGRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243369.1 | NP_001230298.1 | 3 - 4 | 463 | 72 | |
NM_001243370.1 | NP_001230299.1 | 3 - 4 | 462 | 72 | ||
XM_005657038.3 | XP_005657095.1 | 622 | 72 | |||
GenBank mRNA | AY609564.1 | - | 3 - 4 | 447 | 72 | |
CK455001.1 | - | 3 - 4 | 462 | 72 | ||
FD587652.1 | - | 3 - 4 | 471 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
