Assay Details
| Gene Symbol: IGSF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243387.1 | NP_001230316.1 | 1 - 2 | 116 | 71 | |
| GenBank mRNA | AK231345.1 | - | 1 - 2 | 116 | 71 | |
More Information
Set Membership: |
|

IGSF6
Genomic Map