Assay Details
Gene Symbol: BOLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243396.1 | NP_001230325.1 | 1 - 2 | 90 | 82 | |
GenBank mRNA | AK237584.1 | - | 1 - 2 | 79 | 82 | |
AK347441.1 | - | 1 - 2 | 73 | 82 | ||
AK392646.1 | - | 1 - 2 | 110 | 82 | ||
AY609813.1 | - | 1 - 2 | 59 | 82 | ||
FS670697.1 | - | 1 - 2 | 110 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001243396.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
