Assay Details
Gene Symbol: TMX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243407.1 | NP_001230336.1 | 3 - 4 | 487 | 59 | |
GenBank mRNA | AK349470.1 | - | 3 - 4 | 487 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
