Assay Details
| Gene Symbol: HDDC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243409.1 | NP_001230338.1 | 1 - 2 | 128 | 79 | |
| XM_021098000.1 | XP_020953659.1 | 151 | 79 | |||
| GenBank mRNA | AK395620.1 | - | 1 - 2 | 128 | 79 | |
| FS689498.1 | - | 1 - 2 | 128 | 79 | ||
More Information
Set Membership: |
|

HDDC3
Genomic Map