Assay Details
| Gene Symbol: SNRPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243426.1 | NP_001230355.1 | 3 - 4 | 734 | 108 | |
| XM_005670875.3 | XP_005670932.1 | 1098 | 108 | |||
| XM_013983106.2 | XP_013838560.1 | 464 | 108 | |||
| GenBank mRNA | AJ660154.1 | - | 2 - 3 | 225 | 108 | |
| AK348676.1 | - | 3 - 4 | 432 | 108 | ||
| AK400906.1 | - | 3 - 4 | 842 | 108 | ||
More Information
Set Membership: |
|

SNRPD3
Genomic Map