Assay Details
| Gene Symbol: TMIGD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243436.2 | NP_001230365.1 | 2 - 3 | 173 | 99 | |
| GenBank mRNA | CV872063.1 | - | 2 - 3 | 173 | 99 | |
| CX062116.1 | - | 2 - 3 | 181 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001243436.2 transcripts.
Set Membership: |
|

TMIGD3
Genomic Map