Assay Details
| Gene Symbol: NCEH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243484.1 | NP_001230413.1 | 4 - 5 | 725 | 89 | |
| GenBank mRNA | AK344477.1 | - | 4 - 5 | 725 | 89 | |
More Information
Set Membership: |
|

NCEH1
Genomic Map