Assay Details
| Gene Symbol: GNB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243493.1 | NP_001230422.1 | 8 - 9 | 825 | 66 | |
| XM_013987271.2 | XP_013842725.1 | 1784 | 66 | |||
| XM_013987274.1 | XP_013842728.1 | 1176 | 66 | |||
| XM_013987277.1 | XP_013842731.1 | 1769 | 66 | |||
| XM_013987280.1 | XP_013842734.1 | 1292 | 66 | |||
| GenBank mRNA | AK349471.1 | - | 8 - 9 | 825 | 66 | |
More Information
Set Membership: |
|

GNB3
Genomic Map