Assay Details
Gene Symbol: RTRAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243494.1 | NP_001230423.1 | 4 - 5 | 462 | 70 | |
GenBank mRNA | AK240252.1 | - | 4 - 5 | 637 | 70 | |
AK398046.1 | - | 4 - 5 | 648 | 70 | ||
AK398721.1 | - | 4 - 5 | 380 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
