Assay Details
| Gene Symbol: GUCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243502.1 | NP_001230431.1 | 3 - 4 | 411 | 64 | |
| XM_005670881.3 | XP_005670938.1 | 3171 | 64 | |||
| XM_013983107.2 | XP_013838561.1 | 462 | 64 | |||
| GenBank mRNA | AK346425.1 | - | 3 - 4 | 434 | 64 | |
More Information
Set Membership: |
|

GUCD1
Genomic Map