Assay Details
Gene Symbol: SPRYD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243529.1 | NP_001230458.1 | 2 - 3 | 252 | 80 | |
GenBank mRNA | AK392290.1 | - | 2 - 3 | 426 | 80 | |
AY609580.1 | - | 2 - 3 | 252 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
