Assay Details
Gene Symbol: DUOXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243550.1 | NP_001230479.1 | 5 - 6 | 1023 | 84 | |
GenBank mRNA | AK231603.1 | - | 5 - 6 | 1020 | 84 | |
AK345742.1 | - | 5 - 6 | 685 | 84 | ||
DB814958.1 | - | 1 - 2 | 431 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
