Assay Details
Gene Symbol: SEC22C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243553.1 | NP_001230482.1 | 4 - 5 | 711 | 55 | |
XM_005669387.3 | XP_005669444.1 | 1237 | 55 | |||
XM_005669390.3 | XP_005669447.1 | 633 | 55 | |||
XM_005669393.3 | XP_005669450.1 | 675 | 55 | |||
XM_013981500.2 | XP_013836954.1 | 1056 | 55 | |||
XM_013981501.2 | XP_013836955.1 | 1029 | 55 | |||
XM_013981502.2 | XP_013836956.1 | 965 | 55 | |||
XM_021068234.1 | XP_020923893.1 | 1241 | 55 | |||
XM_021068235.1 | XP_020923894.1 | 1034 | 55 | |||
XM_021068236.1 | XP_020923895.1 | 1080 | 55 | |||
XM_021068237.1 | XP_020923896.1 | 1000 | 55 | |||
XM_021068238.1 | XP_020923897.1 | 1039 | 55 | |||
XM_021068239.1 | XP_020923898.1 | 881 | 55 | |||
XM_021068240.1 | XP_020923899.1 | 882 | 55 | |||
XM_021068241.1 | XP_020923900.1 | 880 | 55 | |||
GenBank mRNA | AK347749.1 | - | 4 - 5 | 714 | 55 | |
AK347998.1 | - | 4 - 5 | 638 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
