Assay Details
Gene Symbol: PHB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243556.1 | NP_001230485.1 | 3 - 4 | 492 | 71 | |
GenBank mRNA | AK232271.1 | - | 3 - 4 | 490 | 71 | |
AK393221.1 | - | 3 - 4 | 500 | 71 | ||
AK397943.1 | - | 3 - 4 | 478 | 71 | ||
AK399177.1 | - | 3 - 4 | 411 | 71 | ||
AK399214.1 | - | 3 - 4 | 417 | 71 | ||
AK400998.1 | - | 3 - 4 | 428 | 71 | ||
FS645470.1 | - | 3 - 4 | 414 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
