Assay Details
| Gene Symbol: KHDRBS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243596.1 | NP_001230525.1 | 2 - 3 | 612 | 101 | |
| GenBank mRNA | AK395702.1 | - | 2 - 3 | 646 | 101 | |
| AY609972.1 | - | 2 - 3 | 612 | 101 | ||
More Information
Set Membership: |
|

KHDRBS1
Genomic Map