Assay Details
| Gene Symbol: LHPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243608.1 | NP_001230537.1 | 4 - 5 | 564 | 58 | |
| XM_021071963.1 | XP_020927622.1 | 606 | 58 | |||
| GenBank mRNA | FS690453.1 | - | 4 - 5 | 556 | 58 | |
More Information
Set Membership: |
|

LHPP
Genomic Map