Assay Details
| Gene Symbol: BPNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243632.1 | NP_001230561.1 | 4 - 5 | 1079 | 70 | |
| GenBank mRNA | AK347925.1 | - | 4 - 5 | 1079 | 70 | |
| AK395708.1 | - | 4 - 5 | 1079 | 70 | ||
More Information
Set Membership: |
|

BPNT2
Genomic Map