Assay Details
Gene Symbol: FRA10AC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243654.1 | NP_001230583.1 | 6 - 7 | 517 | 69 | |
XM_005671290.3 | XP_005671347.1 | 581 | 69 | |||
XM_005671293.3 | XP_005671350.1 | 609 | 69 | |||
XM_005671294.3 | XP_005671351.1 | 546 | 69 | |||
XM_013983566.2 | XP_013839020.1 | 433 | 69 | |||
XM_013983569.2 | XP_013839023.1 | 419 | 69 | |||
XM_021072007.1 | XP_020927666.1 | 581 | 69 | |||
GenBank mRNA | AK235945.1 | - | 6 - 7 | 517 | 69 | |
AK346001.1 | - | 7 - 8 | 545 | 69 | ||
AK347603.1 | - | 6 - 7 | 482 | 69 | ||
AK390487.1 | - | 6 - 7 | 529 | 69 | ||
AY610209.1 | - | 6 - 7 | 471 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
