Assay Details
| Gene Symbol: HIST1H2BD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243656.1 | NP_001230585.1 | 1 - 2 | 437 | 67 | |
| GenBank mRNA | DN119932.1 | - | 1 - 2 | 415 | 67 | |
More Information
Set Membership: |
|

HIST1H2BD
Genomic Map