Assay Details
| Gene Symbol: PSIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243670.1 | NP_001230599.1 | 2 - 3 | 391 | 71 | |
| XM_005660092.3 | XP_005660149.1 | 641 | 71 | |||
| XM_021102150.1 | XP_020957809.1 | 641 | 71 | |||
| XM_021102208.1 | XP_020957867.1 | 641 | 71 | |||
| GenBank mRNA | AK349973.1 | - | 2 - 3 | 364 | 71 | |
| AK390475.1 | - | 2 - 3 | 646 | 71 | ||
| FS673276.1 | - | 2 - 3 | 391 | 71 | ||
More Information
Set Membership: |
|

PSIP1
Genomic Map