Assay Details
Gene Symbol: DNTTIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243674.1 | NP_001230603.1 | 1 - 2 | 81 | 66 | |
GenBank mRNA | AK344464.1 | - | 1 - 2 | 95 | 66 | |
AK389813.1 | - | 1 - 2 | 81 | 66 | ||
BW960786.1 | - | 1 - 2 | 81 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
