Assay Details
Gene Symbol: DNTTIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243674.1 | NP_001230603.1 | 5 - 6 | 2131 | 63 | |
GenBank mRNA | AK344464.1 | - | 5 - 6 | 2145 | 63 | |
CF368239.1 | - | 2 - 3 | 403 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
