Assay Details
Gene Symbol: DPH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243679.1 | NP_001230608.1 | 7 - 8 | 789 | 70 | |
XM_005655461.3 | XP_005655518.1 | 800 | 70 | |||
GenBank mRNA | AK344470.1 | - | 7 - 8 | 789 | 70 | |
CK453431.1 | - | 1 - 2 | 37 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
