Assay Details
Gene Symbol: TCEA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243680.1 | NP_001230609.1 | 9 - 10 | 1020 | 68 | |
XM_005653062.3 | XP_005653119.1 | 1088 | 68 | |||
GenBank mRNA | AK349980.1 | - | 9 - 10 | 1020 | 68 | |
CN162678.1 | - | 1 - 2 | 719 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
