Assay Details
Gene Symbol: THOC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243684.1 | NP_001230613.1 | 2 - 3 | 453 | 61 | |
XM_005662202.3 | XP_005662259.1 | 278 | 61 | |||
GenBank mRNA | AK351693.1 | - | 2 - 3 | 453 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
