Assay Details
Gene Symbol: STN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243685.1 | NP_001230614.1 | 4 - 5 | 451 | 77 | |
XM_005671431.3 | XP_005671488.1 | 860 | 77 | |||
XM_005671432.3 | XP_005671489.1 | 858 | 77 | |||
XM_013983705.2 | XP_013839159.1 | 862 | 77 | |||
XM_021072021.1 | XP_020927680.1 | 862 | 77 | |||
XM_021072022.1 | XP_020927681.1 | 861 | 77 | |||
XM_021072023.1 | XP_020927682.1 | 532 | 77 | |||
XM_021072024.1 | XP_020927683.1 | 693 | 77 | |||
XM_021072025.1 | XP_020927684.1 | 691 | 77 | |||
XM_021072027.1 | XP_020927686.1 | 527 | 77 | |||
XM_021072028.1 | XP_020927687.1 | 688 | 77 | |||
GenBank mRNA | AK352421.1 | - | 4 - 5 | 451 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
