Assay Details
| Gene Symbol: TXN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243705.1 | NP_001230634.1 | 2 - 3 | 451 | 80 | |
| XM_005663827.3 | XP_005663884.2 | 338 | 80 | |||
| XM_021090965.1 | XP_020946624.1 | 336 | 80 | |||
| GenBank mRNA | AK236637.1 | - | 2 - 3 | 327 | 80 | |
| AK345890.1 | - | 2 - 3 | 296 | 80 | ||
| AK393937.1 | - | 2 - 3 | 343 | 80 | ||
| AK397007.1 | - | 2 - 3 | 316 | 80 | ||
| AK398328.1 | - | 2 - 3 | 287 | 80 | ||
| AY610430.1 | - | 2 - 3 | 303 | 80 | ||
| CJ013194.1 | - | 2 - 3 | 451 | 80 | ||
| CK454960.1 | - | 1 - 2 | 87 | 80 | ||
More Information
Set Membership: |
|

TXN2
Genomic Map