Assay Details
Gene Symbol: TXN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243705.1 | NP_001230634.1 | 3 - 4 | 578 | 75 | |
XM_005663827.3 | XP_005663884.2 | 465 | 75 | |||
GenBank mRNA | AK236637.1 | - | 3 - 4 | 454 | 75 | |
AK345890.1 | - | 3 - 4 | 423 | 75 | ||
AK393937.1 | - | 3 - 4 | 470 | 75 | ||
AK398328.1 | - | 3 - 4 | 414 | 75 | ||
AY610430.1 | - | 3 - 4 | 430 | 75 | ||
CJ013194.1 | - | 3 - 4 | 578 | 75 | ||
CK454960.1 | - | 2 - 3 | 214 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
