Assay Details
| Gene Symbol: PPP1R14D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243710.1 | NP_001230639.1 | 2 - 3 | 427 | 91 | |
| XM_005659736.3 | XP_005659793.1 | 476 | 91 | |||
| XM_021062777.1 | XP_020918436.1 | 479 | 91 | |||
| XM_021062855.1 | XP_020918514.1 | 478 | 91 | |||
| XM_021062908.1 | XP_020918567.1 | 1271 | 91 | |||
| GenBank mRNA | AY610133.1 | - | 2 - 3 | 427 | 91 | |
More Information
Set Membership: |
|

PPP1R14D
Genomic Map