Assay Details
| Gene Symbol: PRPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243801.1 | NP_001230730.1 | 2 - 3 | 426 | 65 | |
| GenBank mRNA | AK239604.1 | - | 2 - 3 | 426 | 65 | |
| AK394777.1 | - | 1 - 2 | 86 | 65 | ||
| AY609594.1 | - | 2 - 3 | 385 | 65 | ||
| KU705637.1 | - | 2 - 3 | 425 | 65 | ||
More Information
Set Membership: |
|

PRPS2
Genomic Map