Assay Details
| Gene Symbol: CRACR2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243838.1 | NP_001230767.1 | 4 - 5 | 548 | 87 | |
| XM_013989677.2 | XP_013845131.1 | 1226 | 87 | |||
| XM_013989679.2 | XP_013845133.1 | 1190 | 87 | |||
| XM_013989682.2 | XP_013845136.1 | 1226 | 87 | |||
| XM_013989684.2 | XP_013845138.1 | 1318 | 87 | |||
| GenBank mRNA | AK350005.1 | - | 4 - 5 | 548 | 87 | |
More Information
Set Membership: |
|

CRACR2B
Genomic Map