Assay Details
Gene Symbol: SLC25A15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243910.1 | NP_001230839.1 | 4 - 5 | 781 | 63 | |
XM_013980468.2 | XP_013835922.1 | 927 | 63 | |||
XM_021065130.1 | XP_020920789.1 | 539 | 63 | |||
GenBank mRNA | AK394071.1 | - | 4 - 5 | 572 | 63 | |
CV872939.1 | - | 4 - 5 | 560 | 63 | ||
DN118723.1 | - | 1 - 2 | 49 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
