Assay Details
| Gene Symbol: ERGIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243932.1 | NP_001230861.1 | 11 - 12 | 943 | 59 | |
| XM_005655651.3 | XP_005655708.1 | 1018 | 59 | |||
| GenBank mRNA | AK230617.1 | - | 11 - 12 | 943 | 59 | |
More Information
Set Membership: |
|

ERGIC2
Genomic Map