Assay Details
| Gene Symbol: ADH1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243939.1 | NP_001230868.1 | 7 - 8 | 1030 | 101 | |
| GenBank mRNA | AK232739.1 | - | 7 - 8 | 1030 | 101 | |
| AK233345.1 | - | 7 - 8 | 1030 | 101 | ||
| AK233380.1 | - | 7 - 8 | 101 | |||
| AK345798.1 | - | 7 - 8 | 853 | 101 | ||
| AK346112.1 | - | 7 - 8 | 1030 | 101 | ||
| AK350890.1 | - | 7 - 8 | 1052 | 101 | ||
| AK394189.1 | - | 7 - 8 | 1041 | 101 | ||
| AK398005.1 | - | 7 - 8 | 1052 | 101 | ||
More Information
Set Membership: |
|

ADH1C
Genomic Map