Assay Details
| Gene Symbol: MARVELD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243948.1 | NP_001230877.1 | 4 - 5 | 1713 | 77 | |
| XM_021076551.1 | XP_020932210.1 | 1786 | 77 | |||
| XM_021076552.1 | XP_020932211.1 | 1762 | 77 | |||
| XM_021076553.1 | XP_020932212.1 | 1656 | 77 | |||
| XM_021076554.1 | XP_020932213.1 | 1747 | 77 | |||
| GenBank mRNA | AK345912.1 | - | 2 - 3 | 1074 | 77 | |
| AK396289.1 | - | 4 - 5 | 1688 | 77 | ||
More Information
Set Membership: |
|

MARVELD2
Genomic Map