Assay Details
| Gene Symbol: SYNGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244066.1 | NP_001230995.1 | 2 - 3 | 356 | 77 | |
| XM_021091099.1 | XP_020946758.1 | 432 | 77 | |||
| XM_021091101.1 | XP_020946760.1 | 432 | 77 | |||
| XM_021091102.1 | XP_020946761.1 | 432 | 77 | |||
| XM_021091103.1 | XP_020946762.1 | 432 | 77 | |||
| GenBank mRNA | FS667058.1 | - | 2 - 3 | 356 | 77 | |
More Information
Set Membership: |
|

SYNGR1
Genomic Map