Assay Details
| Gene Symbol: ZXDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244222.1 | NP_001231151.1 | 1 - 2 | 905 | 58 | |
| XM_013996783.2 | XP_013852237.1 | 966 | 58 | |||
More Information
Set Membership: |
|

ZXDC
Genomic Map