Assay Details
Gene Symbol: ALG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244234.1 | NP_001231163.1 | 1 - 2 | 320 | 82 | |
GenBank mRNA | AK235622.1 | - | 1 - 2 | 320 | 82 | |
AK236309.1 | - | 1 - 2 | 319 | 82 | ||
BW957963.1 | - | 1 - 2 | 295 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
