Assay Details
| Gene Symbol: SH3BGR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244236.1 | NP_001231165.1 | 7 - 8 | 818 | 62 | |
| GenBank mRNA | AK235947.1 | - | 7 - 8 | 818 | 62 | |
More Information
Set Membership: |
|

SH3BGR
Genomic Map