Assay Details
| Gene Symbol: EFEMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244261.1 | NP_001231190.1 | 4 - 5 | 653 | 64 | |
| XM_013996146.2 | XP_013851600.1 | 649 | 64 | |||
| GenBank mRNA | FD638968.1 | - | 4 - 5 | 611 | 64 | |
| HQ630067.1 | - | 4 - 5 | 673 | 64 | ||
More Information
Set Membership: |
|

EFEMP1
Genomic Map