Assay Details
Gene Symbol: CARD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244277.1 | NP_001231206.1 | 2 - 3 | 264 | 92 | |
NM_001244279.1 | NP_001231208.1 | 2 - 3 | 251 | 92 | ||
GenBank mRNA | AK346684.1 | - | 2 - 3 | 264 | 92 | |
DN114326.1 | - | 2 - 3 | 216 | 92 | ||
EW082630.2 | - | 2 - 3 | 251 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
