Assay Details
Gene Symbol: CARD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244277.1 | NP_001231206.1 | 5 - 6 | 555 | 98 | |
NM_001244279.1 | NP_001231208.1 | 4 - 5 | 482 | 98 | ||
GenBank mRNA | AK346684.1 | - | 5 - 6 | 555 | 98 | |
DN114326.1 | - | 5 - 6 | 507 | 98 | ||
EW082630.2 | - | 4 - 5 | 482 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
