Assay Details
Gene Symbol: ALG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244320.2 | NP_001231249.1 | 3 - 4 | 304 | 66 | |
GenBank mRNA | AK233293.1 | - | 3 - 4 | 312 | 66 | |
AK235803.1 | - | 3 - 4 | 256 | 66 | ||
AK238941.1 | - | 3 - 4 | 314 | 66 | ||
AK239382.1 | - | 3 - 4 | 335 | 66 | ||
AK390932.1 | - | 3 - 4 | 336 | 66 | ||
AY610080.1 | - | 3 - 4 | 325 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
