Assay Details
Gene Symbol: TMEM171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244323.1 | NP_001231252.1 | 2 - 3 | 771 | 88 | |
XM_005661477.2 | XP_005661534.1 | 826 | 88 | |||
XM_013994887.2 | XP_013850341.1 | 909 | 88 | |||
GenBank mRNA | AK345063.1 | - | 2 - 3 | 771 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
