Assay Details
Gene Symbol: HINT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244325.1 | NP_001231254.1 | 1 - 2 | 104 | 99 | |
NM_001244328.1 | NP_001231257.1 | 1 - 2 | 104 | 99 | ||
XM_005660205.3 | XP_005660262.2 | 127 | 99 | |||
GenBank mRNA | AK346311.1 | - | 1 - 2 | 101 | 99 | |
AK392440.1 | - | 1 - 2 | 91 | 99 | ||
AY609781.1 | - | 1 - 2 | 91 | 99 | ||
EV934255.2 | - | 1 - 2 | 81 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
