Assay Details
Gene Symbol: HINT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244325.1 | NP_001231254.1 | 4 - 5 | 426 | 64 | |
NM_001244328.1 | NP_001231257.1 | 4 - 5 | 396 | 64 | ||
XM_005660205.3 | XP_005660262.2 | 344 | 64 | |||
XM_005660206.3 | XP_005660263.1 | 894 | 64 | |||
GenBank mRNA | AK346311.1 | - | 4 - 5 | 423 | 64 | |
AK392440.1 | - | 4 - 5 | 413 | 64 | ||
AY609781.1 | - | 4 - 5 | 413 | 64 | ||
EV934255.2 | - | 4 - 5 | 373 | 64 | ||
EW338246.2 | - | 4 - 5 | 428 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
